3-Amino-2-methoxypyridin

Lieferant: Thermo Scientific

Synonyme: 2-Methoxy-3-pyridylamin, 2-Methoxypyridin-3-amin

SPB07839DA SPB07839EA
MAYBSPB07839DAEA 0 EUR
MAYBSPB07839DA MAYBSPB07839EA
3-Amino-2-methoxypyridin
3-Amino-2-methoxypyridine
Formel: C₆H₈N₂O
Molecular Weight: 124,14 g/mol
MDL: MFCD00833386
CAS-Nummer: 20265-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR