O-(4-Nitrobenzyl)hydroxylamin Hydrochlorid
Lieferant: BIONET RESEARCH
Synonyme:
O-(4-Nitrobenzyl)-hydroxylammonium chloride
7L-014.500
7L-014.1
7L-014.5
7L-014.10
BIOT7L-014.500EA
0
EUR
BIOT7L-014.500
BIOT7L-014.1
BIOT7L-014.5
BIOT7L-014.10
O-(4-Nitrobenzyl)hydroxylamin Hydrochlorid
O-(4-Nitrobenzyl)hydroxylamin Hydrochlorid
Formel:
C₇H₉ClN₂O₃ Molecular Weight: 204.61 g/mol |
MDL:
MFCD00012954 CAS-Nummer: 2086-26-2 EINECS: 218-228-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...