4'-(2-Fluorphenyl)propiophenon

Lieferant: Thermo Scientific

Synonyme: 1-(2'-Fluorobiphenyl-4-yl)propan-1-one

SB01820DA SB01820EA
MAYBSB01820DAEA 0 EUR
MAYBSB01820DA MAYBSB01820EA
4'-(2-Fluorphenyl)propiophenon
4'-(2-Fluorphenyl)propiophenon
Formel: C₁₅H₁₃FO
MDL: MFCD00236612
CAS-Nummer: 37989-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR