N-Boc-4-fluor-3-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

AN-1374-1G AN-1374-25G AN-1374-5G
COBBAN-1374-1GEA 74.2 EUR
COBBAN-1374-1G COBBAN-1374-25G COBBAN-1374-5G
N-Boc-4-fluor-3-(trifluormethyl)anilin
N-Boc-4-fluor-3-(trifluormethyl)anilin
Formel: C₁₂H₁₃F₄NO₂
MDL: MFCD11504831
CAS-Nummer: 1072945-57-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR