1,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(1,5-Dimethyl-1H-pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,5-Dimethyl-1H-pyrazole-4-boronic acid Pinacol ester

PN-8818-1G PN-8818-250M
COBBPN-8818-1GEA 259 EUR
COBBPN-8818-1G COBBPN-8818-250M
1,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
1,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222.0918 g/mol
MDL: MFCD09864189
CAS-Nummer: 1036991-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR