1,5-Dimethyl-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1,5-Dimethyl-1H-pyrazol-4-yl)boronic acid

BB-8818-1G BB-8818-250M
COBBBB-8818-1GEA 1090 EUR
COBBBB-8818-1G COBBBB-8818-250M
1,5-Dimethyl-1H-pyrazol-4-boronsäure
1,5-Dimethyl-1H-pyrazole-4-boronic acid
Formel: C₅H₉BN₂O₂
Molecular Weight: 139,9482 g/mol
MDL: MFCD13195678
CAS-Nummer: 1204333-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR