5-Brom-2-methoxyanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1341-25G AN-1341-1G AN-1341-5G
COBBAN-1341-25GEA 0 EUR
COBBAN-1341-25G COBBAN-1341-1G COBBAN-1341-5G
5-Brom-2-methoxyanilin Hydrochlorid
5-Brom-2-methoxyanilin Hydrochlorid
Formel: C₇H₉BrClNO
MDL: MFCD10699607
CAS-Nummer: 1072945-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR