3-Fluor-2,4-dimethoxyanilin

Lieferant: COMBI-BLOCKS

Synonyme: 2,4-Dimethoxy-3-fluoroaniline

AN-1120-5G AN-1120-1G
COBBAN-1120-5GEA 1350 EUR
COBBAN-1120-5G COBBAN-1120-1G
3-Fluor-2,4-dimethoxyanilin
3-Fluor-2,4-dimethoxyanilin
Formel: C₈H₁₀FNO₂
MDL: MFCD09258660
CAS-Nummer: 195136-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR