Benzyl-(R)-(-)-glycidylether ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-2-((Benzyloxy)methyl)oxirane

L13980.03 L13980.06
L13980.03EA 44.8 EUR
L13980.03 L13980.06
Benzyl-(R)-(-)-glycidylether ≥98%
Benzyl-(R)-(-)-glycidylether

Purity: 98+% CAS No.: 14618-80-5 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 130°/0.1mm Beilstein Reference: 3588399 Optical Rotation: -5.1° (c=5 in toluene) MDL No.: MFCD00068409

Formel: C₁₀H₁₂O₂
Molecular Weight: 164.20 g/mol
Siedepunkt: 130 °C (0.1 mmHg)
Dichte: 1.072
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00068409
CAS-Nummer: 14618-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR