4-(1-Naphthyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(1-Naphthyl)benzeneboronic acid

BB-8826-5G BB-8826-1G
COBBBB-8826-5GEA 113 EUR
COBBBB-8826-5G COBBBB-8826-1G
4-(1-Naphthyl)phenylboronsäure
4-(1-Naphthyl)phenylboronsäure
Formel: C₁₆H₁₃BO₂
Molecular Weight: 248,09 g/mol
MDL: MFCD08669639
CAS-Nummer: 870774-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR