3-Pyrid-3-ylbenzoesäure
Lieferant: FLUOROCHEM
Synonyme:
3-Pyridin-3-yl-benzoic acid
011854-500MG
011854-1G
FLUO011854-500MGEA
0
EUR
FLUO011854-500MG
FLUO011854-1G
3-Pyrid-3-ylbenzoesäure
3-Pyrid-3-ylbenzoesäure
Formel:
C₁₂H₉NO₂ Molecular Weight: 199,2054 g/mol |
MDL:
MFCD03426512 CAS-Nummer: 4385-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...