1,4-Dioxaspiro[4.5]decan-8-on ≥97%
Lieferant: Thermo Scientific
Synonyme: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal
A10408.06 A10408.14 A10408.22
A10408.06EA 36.3 EUR
A10408.06 A10408.14 A10408.22
1,4-Dioxaspiro[4.5]decan-8-on ≥97%
1,4-Dioxaspiro[4.5]decan-8-on
Purity: 97% CAS No.: 4746-97-8 Molecular Formula: C8H12O3 Formula Weight: 156.18 Melting Point: 72-75° Beilstein Reference: 117631
Formel: C₈H₁₂O₃
Molecular Weight: 156,18 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010214
CAS-Nummer: 4746-97-8