1,4-Dioxaspiro[4.5]decan-8-on

Lieferant: COMBI-BLOCKS

Synonyme: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

OT-0403-25G OT-0403-100G OT-0403-5G
COBBOT-0403-25GEA 50.9 EUR
COBBOT-0403-25G COBBOT-0403-100G COBBOT-0403-5G
1,4-Dioxaspiro[4.5]decan-8-on
1,4-Dioxaspiro[4.5]decan-8-on
Formel: C₈H₁₂O₃
Molecular Weight: 156,18 g/mol
MDL: MFCD00010214
CAS-Nummer: 4746-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR