(Chlormethylen)dimethylammoniumchlorid 96%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: N-Chloromethylen-N,N-Dimethyl Ammonium Chloride
B24172.06 B24172.14 B24172.22
B24172.06EA 33 EUR
B24172.06 B24172.14 B24172.22
(Chlormethylen)dimethylammoniumchlorid 96%
(Chlormethylen)dimethylammoniumchlorid
Purity: 96% CAS No.: 3724-43-4 Molecular Formula: C3H7Cl2N Formula Weight: 128.00 Storage Temperature: 2°C to 8°C Odour: Acrid Beilstein Reference: 3566322 UN No.: 3261 MDL No.: MFCD00011868
Formel: C₃H₇Cl₂N
Molecular Weight: 128.00 g/mol
Schmelzpunkt: 130 °C (Zers.)
MDL: MFCD00011868
CAS-Nummer: 3724-43-4
EINECS: 425-970-6
ADR: 8 II