6-Chlorimidazo[1,2-b]pyridazin

Lieferant: COMBI-BLOCKS

HI-1276-5G HI-1276-25G HI-1276-1G
COBBHI-1276-5GEA 38.2 EUR
COBBHI-1276-5G COBBHI-1276-25G COBBHI-1276-1G
6-Chlorimidazo[1,2-b]pyridazin
6-Chlorimidazo[1,2-b]pyridazin
Formel: C₆H₄ClN₃
Molecular Weight: 153,5691 g/mol
MDL: MFCD07778345
CAS-Nummer: 6775-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR