3-Bromimidazo[1,2-a]pyridin
Lieferant: COMBI-BLOCKS
HI-1413-5G
HI-1413-1G
COBBHI-1413-5GEA
147
EUR
COBBHI-1413-5G
COBBHI-1413-1G
3-Bromimidazo[1,2-a]pyridin
3-Bromimidazo[1,2-a]pyridin
Formel:
C₇H₅BrN₂ Molecular Weight: 197,0300 g/mol |
MDL:
MFCD00014556 CAS-Nummer: 4926-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...