6-Bromimidazo[1,2-a]pyridin
Lieferant: BIONET RESEARCH
7X-0903.1
7X-0903.10
7X-0903.5
7X-0903.500
BIOT7X-0903.1EA
0
EUR
BIOT7X-0903.1
BIOT7X-0903.10
BIOT7X-0903.5
BIOT7X-0903.500
6-Bromimidazo[1,2-a]pyridin
6-Bromimidazo[1,2-a]pyridin
Formel:
C₇H₅BrN₂ Molecular Weight: 197.03 g/mol |
MDL:
MFCD06496200 CAS-Nummer: 6188-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...