3-Brom-6-chlorimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HI-1404-5G HI-1404-1G
COBBHI-1404-5GEA 0 EUR
COBBHI-1404-5G COBBHI-1404-1G
3-Brom-6-chlorimidazo[1,2-a]pyridin
3-Brom-6-chlorimidazo[1,2-a]pyridin
Formel: C₇H₄BrClN₂
MDL: MFCD08667858
CAS-Nummer: 886371-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR