3-Brom-7-chlorimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2069-5G HC-2069-1G
COBBHC-2069-5GEA 1240 EUR
COBBHC-2069-5G COBBHC-2069-1G
3-Brom-7-chlorimidazo[1,2-a]pyridin
3-Brom-7-chlorimidazo[1,2-a]pyridin
Formel: C₇H₄BrClN₂
Molecular Weight: 231.4800 g/mol
MDL: MFCD11111834
CAS-Nummer: 342613-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR