3,8-Dibrom-6-chlorimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2125-5G HC-2125-1G
COBBHC-2125-5GEA 0 EUR
COBBHC-2125-5G COBBHC-2125-1G
3,8-Dibrom-6-chlorimidazo[1,2-a]pyridin
3,8-Dibrom-6-chlorimidazo[1,2-a]pyridin
Formel: C₇H₃Br₂ClN₂
MDL: MFCD11504899
CAS-Nummer: 1072944-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR