3-Bromimidazo[1,2-a]pyridin-6-carbonsäure

Lieferant: COMBI-BLOCKS

HC-2501-1G HC-2501-5G HC-2501-250M
COBBHC-2501-1GEA 371 EUR
COBBHC-2501-1G COBBHC-2501-5G COBBHC-2501-250M
3-Bromimidazo[1,2-a]pyridin-6-carbonsäure
3-Bromimidazo[1,2-a]pyridin-6-carbonsäure
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241.05 g/mol (anhy)
MDL: MFCD07021511
CAS-Nummer: 886362-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR