6-Chlor-8-fluorimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-6801-5G HC-6801-1G
COBBHC-6801-5GEA 460 EUR
COBBHC-6801-5G COBBHC-6801-1G
6-Chlor-8-fluorimidazo[1,2-a]pyridin
6-Chlor-8-fluorimidazo[1,2-a]pyridin
Formel: C₇H₄ClFN₂
Molecular Weight: 170,5700 g/mol
MDL: MFCD10699670
CAS-Nummer: 1033202-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR