6-Chlorimidazo[1,2-a]pyridin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-6080-25G
HC-6080-1G
HC-6080-5G
COBBHC-6080-25GEA
595
EUR
COBBHC-6080-25G
COBBHC-6080-1G
COBBHC-6080-5G
6-Chlorimidazo[1,2-a]pyridin Hydrochlorid
6-Chlorimidazo[1,2-a]pyridin Hydrochlorid
Formel:
C₇H₆Cl₂N₂ Molecular Weight: 189,0400 g/mol |
MDL:
MFCD09475876 CAS-Nummer: 957035-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...