7-Chlorimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2068-1G HC-2068-5G
COBBHC-2068-1GEA 0 EUR
COBBHC-2068-1G COBBHC-2068-5G
7-Chlorimidazo[1,2-a]pyridin Hydrochlorid
7-Chlorimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₇H₆Cl₂N₂
MDL: MFCD11504887
CAS-Nummer: 1072944-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR