6-Iodimidazo[1,2-a]pyridin

Lieferant: BIONET RESEARCH

7X-0891.10 7X-0891.5 7X-0891.500 7X-0891.1
BIOT7X-0891.10EA 0 EUR
BIOT7X-0891.10 BIOT7X-0891.5 BIOT7X-0891.500 BIOT7X-0891.1
6-Iodimidazo[1,2-a]pyridin
6-Iodimidazo[1,2-a]pyridin
Formel: C₇H₅IN₂
Molecular Weight: 244.0300 g/mol
MDL: MFCD06200985
CAS-Nummer: 426825-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR