6-Brom-7-methylimidazo[1,2-a]pyridin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-6079-5G
HC-6079-1G
COBBHC-6079-5GEA
408
EUR
COBBHC-6079-5G
COBBHC-6079-1G
6-Brom-7-methylimidazo[1,2-a]pyridin Hydrochlorid
6-Brom-7-methylimidazo[1,2-a]pyridin Hydrochlorid
Formel:
C₈H₈BrClN₂ Molecular Weight: 247.5200 g/mol |
MDL:
MFCD09475875 CAS-Nummer: 957035-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...