6-Brom-7-methyl-3-nitroimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2140-5G HC-2140-1G
COBBHC-2140-5GEA 0 EUR
COBBHC-2140-5G COBBHC-2140-1G
6-Brom-7-methyl-3-nitroimidazo[1,2-a]pyridin
6-Brom-7-methyl-3-nitroimidazo[1,2-a]pyridin
Formel: C₈H₆BrN₃O₂
MDL: MFCD11504905
CAS-Nummer: 1072944-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR