2-Methylimidazo[1,2-a]pyridin-3-carbonsäure

Lieferant: BIONET RESEARCH

10D-545.1 10D-545.10 10D-545.5 10D-545.500
BIOT10D-545.1EA 0 EUR
BIOT10D-545.1 BIOT10D-545.10 BIOT10D-545.5 BIOT10D-545.500
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure
Formel: C₉H₈N₂O₂
MDL: MFCD00244615
CAS-Nummer: 21801-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR