2-Methylimidazo[1,2-a]pyridin-3-carbonsäure

Lieferant: CHEMPUR

RH01017.250 RH01017.1
CHMPRH01017.250EA 0 EUR
CHMPRH01017.250 CHMPRH01017.1
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure
Formel: C₉H₈N₂O₂
MDL: MFCD00244615
CAS-Nummer: 21801-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR