6-Methylimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6165-5G HC-6165-25G HC-6165-1G
COBBHC-6165-5GEA 0 EUR
COBBHC-6165-5G COBBHC-6165-25G COBBHC-6165-1G
6-Methylimidazo[1,2-a]pyridin Hydrochlorid
6-Methylimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₈H₉ClN₂
MDL: MFCD09972136
CAS-Nummer: 10518-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR