7-Methyl-3-nitroimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2004-5G HC-2004-1G
COBBHC-2004-5GEA 0 EUR
COBBHC-2004-5G COBBHC-2004-1G
7-Methyl-3-nitroimidazo[1,2-a]pyridin
7-Methyl-3-nitroimidazo[1,2-a]pyridin
Formel: C₈H₇N₃O₂
MDL: MFCD00179132
CAS-Nummer: 34165-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR