5-Methylimidazo[1,2-a]pyridin
Lieferant: BIONET RESEARCH
8X-0855.1
8X-0855.10
8X-0855.5
8X-0855.500
BIOT8X-0855.1EA
0
EUR
BIOT8X-0855.1
BIOT8X-0855.10
BIOT8X-0855.5
BIOT8X-0855.500
5-Methylimidazo[1,2-a]pyridin
5-Methylimidazo[1,2-a]pyridin
Formel:
C₈H₈N₂ |
MDL:
MFCD06200991 CAS-Nummer: 933-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...