6-Methylimidazo[1,2-a]pyridin
Lieferant: BIONET RESEARCH
7X-0847.500
7X-0847.1
7X-0847.5
7X-0847.10
BIOT7X-0847.500EA
0
EUR
BIOT7X-0847.500
BIOT7X-0847.1
BIOT7X-0847.5
BIOT7X-0847.10
6-Methylimidazo[1,2-a]pyridin
6-Methylimidazo[1,2-a]pyridin
Formel:
C₈H₈N₂ Molecular Weight: 132,1625 g/mol |
MDL:
MFCD05663806 CAS-Nummer: 874-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...