7-Methylimidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-6162-5G HC-6162-25G HC-6162-1G
COBBHC-6162-5GEA 507 EUR
COBBHC-6162-5G COBBHC-6162-25G COBBHC-6162-1G
7-Methylimidazo[1,2-a]pyridin
7-Methylimidazo[1,2-a]pyridin
Formel: C₈H₈N₂
Molecular Weight: 132.1600 g/mol
MDL: MFCD06761695
CAS-Nummer: 874-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR