8-Methylimidazo[1,2-a]pyridin

Lieferant: BIONET RESEARCH

7X-0831.500 7X-0831.1 7X-0831.5 7X-0831.10
BIOT7X-0831.500EA 0 EUR
BIOT7X-0831.500 BIOT7X-0831.1 BIOT7X-0831.5 BIOT7X-0831.10
8-Methylimidazo[1,2-a]pyridin
8-Methylimidazo[1,2-a]pyridin
Formel: C₈H₈N₂
MDL: MFCD05663805
CAS-Nummer: 874-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR