2-(4-Chlorphenyl)imidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2049-5G HC-2049-1G
COBBHC-2049-5GEA 451 EUR
COBBHC-2049-5G COBBHC-2049-1G
2-(4-Chlorphenyl)imidazo[1,2-a]pyridin
2-(4-Chlorphenyl)imidazo[1,2-a]pyridin
Formel: C₁₃H₉ClN₂
Molecular Weight: 228.677 g/mol
MDL: MFCD00185874
CAS-Nummer: 38922-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR