3-Chlor-6-(4-methyl-1-imidazolyl)pyridazin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-6-(4-methyl-1H-imidazol-1-yl)pyridazine

HC-6044-5G HC-6044-1G
COBBHC-6044-5GEA 667 EUR
COBBHC-6044-5G COBBHC-6044-1G
3-Chlor-6-(4-methyl-1-imidazolyl)pyridazin
3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine
Formel: C₈H₇ClN₄
Molecular Weight: 194.6210 g/mol
MDL: MFCD09800928
CAS-Nummer: 941294-26-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR