3-(3,5-Dimethyl-4-ethoxyphenyl)anilin

Lieferant: COMBI-BLOCKS

AN-1396-5G AN-1396-1G
COBBAN-1396-5GEA 451 EUR
COBBAN-1396-5G COBBAN-1396-1G
3-(3,5-Dimethyl-4-ethoxyphenyl)anilin
3-(3,5-Dimethyl-4-ethoxyphenyl)anilin
Formel: C₁₆H₁₉NO
MDL: MFCD12913929
CAS-Nummer: 1187386-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR