1H-Pyrazol-4-boronsäurepinakolester

Lieferant: FLUOROCHEM

010845-5G 010845-1G
FLUO010845-5GEA 76.2 EUR
FLUO010845-5G FLUO010845-1G
1H-Pyrazol-4-boronsäurepinakolester
1H-Pyrazol-4-boronsäurepinakolester
Formel: C₉H₁₅BN₂O₂
MDL: MFCD03453063
CAS-Nummer: 269410-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR