1H-Pyrazole-5-boronic acid

Lieferant: COMBI-BLOCKS

Synonyme: 5-Borono-1H-pyrazole, 2H-Pyrazole-3-boronic acid, (1H-Pyrazol-5-yl)boronic acid

BB-8515-5G BB-8515-1G
COBBBB-8515-5GEA 321 EUR
COBBBB-8515-5G COBBBB-8515-1G
1H-Pyrazole-5-boronic acid
1H-Pyrazole-5-boronic acid
Formel: C₃H₅BN₂O₂
Molecular Weight: 111,89 g/mol
MDL: MFCD07368247
CAS-Nummer: 376584-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR