1H-Pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

BB-8516-5G BB-8516-1G
COBBBB-8516-5GEA 510 EUR
COBBBB-8516-5G COBBBB-8516-1G
1H-Pyrazol-4-boronsäure
1H-Pyrazole-4-boronic acid
Formel: C₃H₅BN₂O₂
Molecular Weight: 111,9000 g/mol
MDL: MFCD07368251
CAS-Nummer: 763120-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR