1H-Pyrazol-4-boronsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
706256-1G
706256-250MG
SIAL706256-1GEA
132
EUR
SIAL706256-1G
SIAL706256-250MG
1H-Pyrazol-4-boronsäure, Sigma-Aldrich®
1H-Pyrazole-4-boronic acid
Formel:
C₃H₅BN₂O₂ Molecular Weight: 111,9000 g/mol |
MDL:
MFCD07368251 CAS-Nummer: 763120-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...