1H-Pyrazol-4-boronsäure Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
(1H-Pyrazol-4-yl)boronic acid hydrochloride
BB-6453-5G
BB-6453-1G
COBBBB-6453-5GEA
486
EUR
COBBBB-6453-5G
COBBBB-6453-1G
1H-Pyrazol-4-boronsäure Hydrochlorid
1H-Pyrazol-4-boronsäure Hydrochlorid
Formel:
C₃H₆BClN₂O₂ Molecular Weight: 148.36 g/mol |
MDL:
MFCD16618935 CAS-Nummer: 1256346-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...