1,3,5-Trimethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1,3,5-Trimethylpyrazole-4-boronic acid Pinacol ester, 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1,3,5-Trimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8661-5G PN-8661-1G PN-8661-250M
COBBPN-8661-5GEA 341 EUR
COBBPN-8661-5G COBBPN-8661-1G COBBPN-8661-250M
1,3,5-Trimethyl-1H-pyrazol-4-boronsäurepinakolester
1,3,5-Trimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₂H₂₁BN₂O₂
Molecular Weight: 236.12 g/mol
MDL: MFCD06659062
CAS-Nummer: 844891-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR