1,3,5-Trimethyl-1H-pyrazol-4-boronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(1,3,5-Trimethyl-1H-pyrazol-4-yl)boronic acid
BB-8661-5G
BB-8661-1G
COBBBB-8661-5GEA
811
EUR
COBBBB-8661-5G
COBBBB-8661-1G
1,3,5-Trimethyl-1H-pyrazol-4-boronsäure
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid
Formel:
C₆H₁₁BN₂O₂ Molecular Weight: 153.9800 g/mol |
MDL:
MFCD11183310 CAS-Nummer: 847818-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...