(S)-2,2'-Dimethoxy-1,1'-binaphthalin-3,3'-diboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2,2'-Dimethoxy-[1,1'-binaphthalene]-3,3'-diyl)diboronic acid, (S)-2,2'-Dimethoxy-1,1'-binaphthyl-3,3'-diboronic acid

BB-3836-1G BB-3836-250M
COBBBB-3836-1GEA 303 EUR
COBBBB-3836-1G COBBBB-3836-250M
(S)-2,2'-Dimethoxy-1,1'-binaphthalin-3,3'-diboronsäure
(S)-2,2'-Dimethoxy-1,1'-binaphthalin-3,3'-diboronsäure
Formel: C₂₂H₂₀B₂O₆
Molecular Weight: 402.02 g/mol
MDL: MFCD09027249
CAS-Nummer: 220204-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR