(S)-2,2'-Dihydroxy-1,1'-binaphthalin-3,3'-diboronsäure

Lieferant: COMBI-BLOCKS

BB-3902-1G BB-3902-250M
COBBBB-3902-1GEA 1630 EUR
COBBBB-3902-1G COBBBB-3902-250M
(S)-2,2'-Dihydroxy-1,1'-binaphthalin-3,3'-diboronsäure
(S)-2,2'-Dihydroxy-1,1'-binaphthalin-3,3'-diboronsäure
Formel: C₂₀H₁₆B₂O₆
MDL: MFCD09258753
CAS-Nummer: 957111-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR