2-Fluor-1-methylpyridinium-p-toluolsulfonat

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate

009219-5G 009219-25G
FLUO009219-5GEA 251 EUR
FLUO009219-5G FLUO009219-25G
2-Fluor-1-methylpyridinium-p-toluolsulfonat
2-Fluor-1-methylpyridinium-p-toluolsulfonat
Formel: C₁₃H₁₄FNO₃S
Molecular Weight: 283,32 g/mol
MDL: MFCD00011983
CAS-Nummer: 58086-67-2
EINECS: 261-108-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR