2-Fluor-1-methylpyridinium-p-toluolsulfonat
Lieferant: CHEMPUR
Synonyme:
2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate
JFD03653.250
JFD03653.1
CHMPJFD03653.250EA
0
EUR
CHMPJFD03653.250
MAYBJFD03653.1
2-Fluor-1-methylpyridinium-p-toluolsulfonat
2-Fluor-1-methylpyridinium-p-toluolsulfonat
Formel:
C₁₃H₁₄FNO₃S Molecular Weight: 283,32 g/mol |
MDL:
MFCD00011983 CAS-Nummer: 58086-67-2 EINECS: 261-108-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...