1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dichloro-1-fluoropyridin-1-ium tetrafluoroborate, 2,6-Dichloro-N-fluoropyridinium tetrafluoroborate

006225-5G 006225-1G
FLUO006225-5GEA 330 EUR
FLUO006225-5G FLUO006225-1G
1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat
1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat
Formel: C₅H₃BCl₂F₅N
Molecular Weight: 253,793 g/mol
MDL: MFCD00153157
CAS-Nummer: 140623-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR